Mouse IgG1 kappa Isotype Control [clone: MOPC-21]
76284-598
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Western Blot:Yes
- Size:200 µg
- Form:Liquid
- Storage Buffer:In PBS containing 0.02% Proclin 300.
- Storage Temperature:–20 °C
- Shipping Temperature:Shipped on Blue Ice
- Purification:Protein A affinity purified.
- Cat. No.:76284-598
Recombinant monclonal antibody (Mouse IgG1κ).
Recommended Applications: Flow Cytometry, WB
Type: Control
Antigen:
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1kappa
Reactivity: