Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fittings and Connectors
HPLC Male Nuts Fittings, M6 Thread, MicroSolv
HPLC Male Nuts Fittings, M6 Thread, MicroSolv
10803-846
:
MicroSolv Technology Corp
HPLC Male Nuts Fittings, M6 Thread, MicroSolv
10803-846
:
MicroSolv Technology Corp
:
47301-M6
Description:
Male nuts fitttings
Length:
15 mm
Material:
PEEK
Cat. No.:
10803-846
For use with:
¹/₁₆" tubing
Supplier no.:
47301-M6