ContactUsLinkComponent
HEPA Filter, Airclean Systems
  MSPP-ACFHEPA
HEPA Filter, Airclean Systems
  MSPP-ACFHEPA
 :  ACFHEPA
  • Description:
    HEPA Filter
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-ACFHEPA
  • Supplier no.:
    ACFHEPA
High Efficiency Particulate Air (HEPA) Filters are available for the containment of particles.
The filter has a 99.997% efficiency rating for collecting 0.3 µm particles.