BluePippin™ Gel Cassette Kit, Sage Science
MSPP-BLF7510
- Description:BluePippin™ Gel Cassette Kit, Dye-Free
- Size:1 - 10 kb
- Cat. No.:MSPP-BLF7510
- Includes:0.75% Dye-Free Agarose Gel Cassette and External Marker S1
- Supplier no.:BLF7510
Cassettes are disposable and pre-cast with agarose and running buffer. Samples are not optically detected.