Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Human IgE, Myeloma
- Size:100 µg
- Cat. No.:80052-344
- Storage temperature:≤ –70 °C
- Supplier no.:401152-100UG
Specifications
About this item
Human IgE, Myeloma is validated for use as an immunoassay standard.
- Formulation: In 150 mM NaCl, 15 mM potassium phosphate buffer, pH 7.4
- Preservative: ≤0.1% sodium azide
- Purity: Single arc by immunoelectrophoresis against antiserum to whole human serum and human IgE
- Source: Prepared from plasma that was shown by certified tests to be nonreactive for HBsAg and for antibodies to HCV, HBc, HIV-1 and HIV-2
The heavy chain of myeloma IgE may appear as either a single or double band on gel electrophoresis. Variables associated with assay conditions will dictate the proper working dilution.