Order Entry
United States
ContactUsLinkComponent
ApoScreen® Human Recombinant Annexin V (from E. coli), Cy5®
ApoScreen® Human Recombinant Annexin V (from E. coli), Cy5®
Catalog # 100242-244
CAS Number:  
ApoScreen® Human Recombinant Annexin V (from E. coli), Cy5®
Catalog # 100242-244
Supplier Number:  10040-15
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Cy5®
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 tests
  • Storage Conditions:
    2-8 °C
  • Gene ID:
    NCBI 308
  • Protein Synonyms:
    ANX5|AXV|Annexin A5
  • Protein/Peptide Name:
    Annexin V
  • Concentration:
    Lot specific
  • Formulation:
    Supplied in ApoScreen® Annexin V Binding Buffer
  • Shipping Temperature:
    Ambient
  • Cat. No.:
    100242-244
  • Supplier no.:
    10040-15

Specifications

About this item

ApoScreen® Annexin V (SB Cat. No. 10040) is a recombinant protein that is expressed in E. coli. A number of changes in cell structure occur during apoptosis. One major change is the loss of membrane phospholipid asymmetry, with translocation of phosphatidylserine (PS) from the inner leaflet of the phospholipid bilayer to the external surface. Annexin V, a member of the annexin family of calcium-dependent phospholipid binding proteins, has a high affinity for phosphatidylserine (PS)-containing phospholipid bilayers. Conjugated Annexin V can be used to monitor changes in cell membrane phospholipid asymmetry, thereby providing a convenient tool for detection of apoptotic cells.