Rat Native Type I Collagen (from Tail tendon)
Supplier: Southern Biotechnology
Certificates
About this item
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Native
- Source:Tail tendon
- Species:Rat
- Storage Conditions:2-8 °C
- Gene ID:NCBI 29393/84352
- Reconstitution Instructions:Reconstitute in 500 mM acetic acid (Note - May be transferred into physiological buffers by dialysis at 2-8°C)
- Protein Synonyms:COL1
- Protein/Peptide Name:Type I Collagen
- Purity:>90% by SDS-PAGE
- Formulation:Supplied lyophilized from 500 mM acetic acid
- Shipping Temperature:4 °C
- Concentration:0.5 mg/mL
Specifications
Frequently Bought Together