Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Secondary
- Antigen Name:Lambda
- Antigen Symbol:Lambda
- Clonality:Polyclonal
- Conjugation:RPE (R-Phycoerythrin)
- Flow Cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Size:0.5 mg
- Cross Adsorption:Cross-adsorbed against human κ light chains for minimal reactivity
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Human λ light chains
- Tested Applications:FLISA
- Purification:Pooled antisera from goats hyperimmunized with human λ light chains; purified by affinity chromatography on human λ light chains covalently linked to agarose; reacts with human λ light chains; cross-adsorbed against human κ light chains for minimal reactivity
- Cat. No.:100244-384
- Supplier no.:2070-09
Specifications
About this item
Goat Anti-Human Lambda-PE antibody for use in flow cytometry assays.
Type: Secondary
Antigen: Lambda
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human