Order Entry
United States
ContactUsLinkComponent
Anti-Kappa Goat Antibody (Alexa Fluor® 555)
Anti-Kappa Goat Antibody (Alexa Fluor® 555)
Catalog # 103259-998
Anti-Kappa Goat Antibody (Alexa Fluor® 555)
Catalog # 103259-998
Supplier Number:  2060-32
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Kappa
  • Antigen Symbol:
    Kappa
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    1.0 mg
  • Environmentally Preferable:
  • Cross Adsorption:
    Cross-adsorbed against human λ light chains for minimal reactivity
  • Form:
    Liquid
  • Storage Buffer:
    Supplied in PBS/NaN3
  • Storage Temperature:
    2-8°C
  • Concentration:
    1.0 mg/mL
  • Shipping Temperature:
    Ambient Temperature
  • Immunogen:
    Human κ light chains
  • Tested Applications:
    FLISA
  • Purification:
    Pooled antisera from goats hyperimmunized with human κ light chains; purified by affinity chromatography on human κ light chains covalently linked to agarose; reacts with human κ light chains; cross-adsorbed against human λ light chains for minimal reactivity
  • Cat. No.:
    103259-998
  • Supplier no.:
    2060-32

Specifications

About this item

Goat Anti-Human Kappa-Alexa Fluor 555 antibody for use in immunohistochemistry / immunocytochemistry and western blotting assays.

Type: Secondary
Antigen: Kappa
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human