Order Entry
United States
ContactUsLinkComponent
Anti-IgG2b Rat Antibody (AP (Alkaline Phosphatase)) [clone: SB74g]
Anti-IgG2b Rat Antibody (AP (Alkaline Phosphatase)) [clone: SB74g]
Catalog # 103259-854
Anti-IgG2b Rat Antibody (AP (Alkaline Phosphatase)) [clone: SB74g]
Catalog # 103259-854
Supplier Number:  1186-04
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G2b
  • Antigen Symbol:
    IgG2b
  • Clonality:
    Monoclonal
  • Clone:
    SB74g
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    1.0 mL
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    16016
  • Storage Buffer:
    Supplied in a stock solution of 50 mM Tris/1 mM MgCl2/50% glycerol, pH 8.0, containing NaN3 as a preservative
  • Storage Temperature:
    2-8°C
  • Concentration:
    Lot specific
  • Shipping Temperature:
    Ambient Temperature
  • Immunogen:
    Mouse IgG2b hybridoma
  • Cat. No.:
    103259-854
  • Supplier no.:
    1186-04

Specifications

About this item

AP Anti-Mouse IgG2b antibody for use in ELISA,, immunocytochemistry, and western blotting assays.

Type: Secondary
Antigen: IgG2b
Clonality: Monoclonal
Clone: SB74g
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse