Order Entry
United States
ContactUsLinkComponent
Anti-Mouse Lambda Rat Antibody [clone: JC5-1]
Anti-Mouse Lambda Rat Antibody [clone: JC5-1]
  100242-868
Anti-Mouse Lambda Rat Antibody [clone: JC5-1]
  100242-868
 :  1175-01

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody Type:
    Secondary
  • Antigen Name:
    Lambda
  • Antigen Symbol:
    Lambda
  • Clonality:
    Monoclonal
  • Clone:
    JC5-1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.5 mg
  • Form:
    Liquid
  • Gene ID:
    111519
  • Storage Buffer:
    Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    Ambient Temperature
  • Tested Applications:
    Sep
  • Cat. No.:
    100242-868
  • Supplier no.:
    1175-01

 

 

Purified Anti-Mouse Lambda antibody for use in ELISA, flow cytometry, immunohistochemistry, western blotting, and separation assays.

Type: Secondary
Antigen: Lambda
Clonality: Monoclonal
Clone: JC5-1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse