Order Entry
United States
ContactUsLinkComponent
Anti-IgG2b Goat Antibody (Biotin)
Anti-IgG2b Goat Antibody (Biotin)
Catalog # 103259-814
Anti-IgG2b Goat Antibody (Biotin)
Catalog # 103259-814
Supplier Number:  1091-08
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G2b
  • Antigen Symbol:
    IgG2b
  • Clonality:
    Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Size:
    1.0 mg
  • Environmentally Preferable:
  • Cross Adsorption:
    Cross-adsorbed against mouse IgG1, IgG2a, IgG3, IgM, and IgA for minimal reactivity
  • Form:
    Liquid
  • Storage Buffer:
    Supplied in PBS/NaN3
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    Ambient Temperature
  • Immunogen:
    Mouse IgG2b
  • Purification:
    Pooled antisera from goats hyperimmunized with mouse IgG2b; purified by affinity chromatography on mouse IgG2b covalently linked to agarose; reacts with the heavy chain of mouse IgG2b; cross-adsorbed against mouse IgG1, IgG2a, IgG3, IgM, and IgA for minimal reactivity
  • Cat. No.:
    103259-814
  • Supplier no.:
    1091-08

Specifications

About this item

Goat Anti-Mouse IgG2b-Biotin antibody for use in ELISA assays.

Type: Secondary
Antigen: IgG2b
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse