Order Entry
United States
ContactUsLinkComponent
Anti-GM-CSF Rat Monoclonal Antibody [clone: MP1-22E9]
Anti-GM-CSF Rat Monoclonal Antibody [clone: MP1-22E9]
Catalog # 100242-466
Anti-GM-CSF Rat Monoclonal Antibody [clone: MP1-22E9]
Catalog # 100242-466
Supplier Number:  10235-01

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    GM-CSF
  • Clonality:
    Monoclonal
  • Clone:
    MP1-22E9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.5 mg
  • Form:
    Liquid
  • Gene ID:
    12981
  • Antigen Synonyms:
    Eo-CSF|CSF-α|pluripoietin-α|eosinophil colony stimulating factor|BPA|Granulocyte/macrophage colony-stimulating factor|burst promoting activity|GM CSF
  • Storage Buffer:
    Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added
  • Storage Temperature:
    2-8°C
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    Ice Packs
  • Immunogen:
    COS-expressed mouse GM-CSF
  • Tested Applications:
    ELISPOT, Multiplex
  • Cat. No.:
    100242-466
  • Supplier no.:
    10235-01

Specifications

About this item

Purified Anti-Mouse GM-CSF antibody for use in flow cytometry, immunohistochemistry / immunocytochemistry, western blotting, ELISA, ELISpot, and multiplex assays.

Type: Primary
Antigen: GM-CSF
Clonality: Monoclonal
Clone: MP1-22E9
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse