To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD90.2
- Antigen Symbol:CD90.2
- Clonality:Monoclonal
- Clone:30-H12
- Conjugation:Pacific Blue™
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:21838
- Antigen Synonyms:Thy-1.2
- Storage Buffer:Supplied in PBS/NaN3
- Storage Temperature:2-8°C
- Concentration:0.5 mg/mL
- Shipping Temperature:Ambient Temperature
- Immunogen:Mouse thymus or spleen
- Cat. No.:103259-952
- Supplier no.:1750-26
Specifications
About this item
CD90 (Thy-1) is a GPI-anchored molecule and one of the smallest members of the immunoglobulin superfamily consisting of a single V-set domain. The Thy-1.2 alloantigen is expressed on all thymocytes, peripheral T lymphocytes, and some intraepithelial T cells of most mouse strains. It is also expressed in the brain and at varying levels on other non-lymphoid tissues. The 30-H12 monoclonal antibody does not cross react with mouse and rat strains bearing the Thy-1.1 alloantigen (e.g., AKR/J, PL).
Type: Primary
Antigen: CD90.2
Clonality: Monoclonal
Clone: 30-H12
Conjugation: Pacific Blue™
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse