To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Type I Collagen
- Antigen Symbol:Type I Collagen
- Clonality:Monoclonal
- Clone:4F6
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:1.0 mL
- Environmentally Preferable:
- Form:Liquid
- Gene ID:1277/1278
- Antigen Synonyms:COL1
- Storage Buffer:Supplied in a stock solution of 50% glycerol/50% PBS, pH 7.4; No preservative added
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Immunogen:Native human type I collagen
- Cat. No.:103264-518
- Supplier no.:1441-05
Specifications
About this item
HRP Anti-Human Type I Collagen antibody for use in ELISA and immunohistochemistry assays.
Type: Primary
Antigen: Type I Collagen
Clonality: Monoclonal
Clone: 4F6
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human