To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Control
- Antigen Name:Keyhole limpet hemocyanin
- Antigen Symbol:KLH
- Clonality:Monoclonal
- Clone:KLH/G2b-1-2
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:
- Size:0.5 mg
- Environmentally Preferable:
- Form:Liquid
- Storage Buffer:Supplied in PBS
- Storage Temperature:–20 °C
- Concentration:0.5 mg/mL
- Shipping Temperature:Dry Ice
- Tested Applications:In vivo Control
- Cat. No.:100242-112
- Supplier no.:0118-14
Specifications
About this item
Low Endotoxin/Azide Free Rat IgG2b for use as a control in flow cytometry, immunohistochemistry, and in vivo assays and as a standard in ELISA.
Type: Control
Antigen: KLH
Clonality: Monoclonal
Clone: KLH/G2b-1-2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: