To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Control
- Antigen Name:T-2 mycotoxin
- Antigen Symbol:T-2 mycotoxin
- Clonality:Monoclonal
- Clone:15H6
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Antibody Fragment:F(ab')2
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Tested Applications:FLISA
- Cat. No.:100242-060
- Supplier no.:0115-11
Specifications
About this item
Mouse F(ab’)2 IgG1-APC for use as a control in flow cytometry assays.
Type: Control
Antigen: T-2 mycotoxin
Clonality: Monoclonal
Clone: 15H6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: