Goat IgG Isotype Control (APC (Allophycocyanin))
Catalog # 100242-024
Supplier: Southern Biotechnology
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Control
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Storage Buffer:Supplied in PBS/NaN3 and a stabilizing agent
- Storage Temperature:2-8°C
- Concentration:Lot specific
- Shipping Temperature:Ambient Temperature
- Tested Applications:FLISA
- Cat. No.:100242-024
- Supplier no.:0109-11
Specifications
About this item
Goat IgG-APC for use as a control in flow cytometry assays.
Type: Control
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: