ContactUsLinkComponent
Magnetic Bead Separation Blocks, 384 Well Microplates, V&P SCIENTIFIC INC MS
Magnetic Bead Separation Blocks, 384 Well Microplates, V&P SCIENTIFIC INC MS
Catalog # MSPP-771G4AAM1
Magnetic Bead Separation Blocks, 384 Well Microplates, V&P SCIENTIFIC INC MS
Catalog # MSPP-771G4AAM1
Supplier Number:  VP771G-4AAZM-1
Specifications
  • Description:
    Magnetic bead separation block for 384 microplates
  • Cat. No.:
    MSPP-771G4AAM1
  • Supplier no.:
    771G-4AAM-1
Specifications
About this item
Designed for robotic gripper access for microplate movement.
  • Red anodized aluminum frame
  • Zinc coated steel plate
  • 425 post 52 MGO neodymium magnet
  • SBS footprint
Magnetic bead separation block is used for 384 well microplates with flat bottom wells only.
Bead pellets are located in the 4 corners/sides, bottom.
North poles up, axial magnetic orientation.
Delivery: Includes anodized aluminum registration base.