ContactUsLinkComponent
DecapiCones® Rodent Restrainer, Braintree Scientific
DecapiCones® Rodent Restrainer, Braintree Scientific
Catalog # MSPP-DCL120
DecapiCones® Rodent Restrainer, Braintree Scientific
Catalog # MSPP-DCL120
Supplier Number:  DC-L120
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    DecapiCones for large rats
  • Cat. No.:
    MSPP-DCL120
  • Packaging:
    120
Specifications
About this item
Tapered plastic film tubes provide quick and easy restraint of rats, mice, and other small animals.
  • Injections can be made directly through the film
  • Restrain post-decapitation kicking
  • Prevent personal contact with feces or urine
A unique dispenser holds Decapicones open and ready for use.
They may be used repeatedly for injections and simply discarded when soiled. For decapitation, hold at the rear and insert the small end into the decapitator.