Safe-Lock Protein LoBind Tubes, Eppendorf®
Supplier: Avantor
Certificates
About this item
Eppendorf LoBind material guarantees maximum sample recovery for improved assay results.
- PCR-clean
- Certified free from DNA, DNase, RNase, and PCR inhibitors
Frequently Bought Together