Westergren Erythrocyte Sedimentation Rate Tubes, Polymedco
Ideal for polymedco sediplast autozero ESR system in westergren erthyrocyte sedimentation.
- Automatic self-zeroing cap and reservoir
- Prefilled diluent and low sample volume
- Pierceable stopper
- Closed system with unique vented cap
Precisely calibrated hole stops the column of blood at true zero with diluent.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.