Specifications
- Description:Single-Stranded DNA Binding Protein
- Size:100 ug
- Cat. No.:99902-310
- Supplier no.:600201
- Details:The single-stranded DNA binding protein is useful in the study of DNA replication, recombination, and repair processes.
Specifications
About this item
The single-stranded DNA binding protein is a helix-destabilizing E.coli protein which is essential for replication, recombination, and repair processes.
- Facilitates sequencing and site-directed mutagenesis reactions
This 75.6 kDa protein consists of four identical monomers and selectively binds single-stranded DNA.