Order Entry
United States
ContactUsLinkComponent
Human Recombinant Tetra-Ubiquitin (from E. coli)
Human Recombinant Tetra-Ubiquitin (from E. coli)
Catalog # 102890-880
Supplier:  Avantor
CAS Number:  
Human Recombinant Tetra-Ubiquitin (from E. coli)
Catalog # 102890-880
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Gene ID:
    7314
  • Protein Synonyms:
    TetraUbiquitin|Ub4|HEL-S-50|UBB|ubiquitin B
  • Accession No.:
    P0CG47
  • Protein/Peptide Name:
    Tetra-Ubiquitin
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain
  • Cat. No.:
    102890-880
  • Supplier no.:
    UCN-710-100

Specifications

About this item

The Recombinant Human Linear Tetra-Ub Non-hydrolyzable from R&D Systems, powered by R&D Systems, powered by Boston Biochem is derived from E. coli. The Recombinant Human Linear Tetra-Ub Non-hydrolyzable has been validated for the following applications: Enzyme Activity.