To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 μg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8 C as supplied. 1 month at 2-8 C under sterile conditions after reconstitution. 3 months at -20 to -80 C under sterile conditions after reconstitution.
- Endotoxin Content:<0.01 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:2247
- Protein Synonyms:Prostatropin|HBGF-2|HBGH-2|FGF2|FGFBprostatropin|bFGF|NUDT6|FGF-2|basic fibroblast growth factor bFGF|Basic fibroblast growth factor|FGF2AS|fibroblast growth factor 2 (basic)|GFG1|heparin-binding growth factor 2
- Accession No.:PO9O38
- Protein/Peptide Name:FGF basic/FGF2
- Purity:>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
- Molecular Weight:
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Cat. No.:102887-858
- Supplier no.:PRD233-10
The ProDots Recombinant Human FGF basic (146 aa) Protein from R&D Systems is derived from E. coli. The ProDots Recombinant Human FGF basic (146 aa) Protein has been validated for the following applications: Bioactivity.