Order Entry
United States
ContactUsLinkComponent
Human Recombinant Ubiquitin-activating Enzyme/UBE1 (from Baculovirus (Sf9 Insect Cells))
Human Recombinant Ubiquitin-activating Enzyme/UBE1 (from Baculovirus (Sf9 Insect Cells))
Catalog # 102880-666
Supplier:  R&D Systems
CAS Number:  
Human Recombinant Ubiquitin-activating Enzyme/UBE1 (from Baculovirus (Sf9 Insect Cells))
Catalog # 102880-666
Supplier:  R&D Systems
Supplier Number:  E-306-050
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Sf9 Insect cells)
  • Species:
    Human
  • Size:
    50 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
  • Gene ID:
    7317
  • Protein Synonyms:
    A1ST|UBA1A|ubiquitin-activating enzyme E1 (A1S9T and BN75 temperature sensitivitycomplementing)|UBE1X|GXP1|ubiquitin-activating enzyme E1 homolog A|A1S9T|ubiquitin-like modifier-activating enzyme 1|UBE1|POC20 centriolar protein homolog|Ubiquitinactivating Enzyme|POC20|AMCX1|A1S9T and BN75 temperature sensitivity complementing|SMAX2|MGC4781|UBA1|Protein A1S9|ubiquitin-like modifier activating enzyme 1|Ubiquitin-activating enzyme E1|UBE1A1S9
  • Accession No.:
    P22314
  • Protein/Peptide Name:
    Ubiquitin-activating Enzyme/UBE1
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain
  • Cat. No.:
    102880-666
  • Supplier no.:
    E-306-050

Specifications

About this item

The Recombinant Human GST-Ubiquitin E1 Enzyme (UBE1) Protein from R&D Systems, powered by R&D Systems, powered by Boston Biochem is derived from Sf 9 (baculovirus). The Recombinant Human GST-Ubiquitin E1 Enzyme (UBE1) Protein has been validated for the following applications: Enzyme Activity.