Order Entry
United States
ContactUsLinkComponent
Human Recombinant GDF-15 (from HEK293 Cells)
Human Recombinant GDF-15 (from HEK293 Cells)
Catalog # 102869-342
Supplier:  R&D Systems
CAS Number:  
Human Recombinant GDF-15 (from HEK293 Cells)
Catalog # 102869-342
Supplier:  R&D Systems
Supplier Number:  8146-GD-025/CF
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    25 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    9518
  • Protein Synonyms:
    growth differentiation factor 15|PLABNRG-1|growth/differentiation factor 15|NAG-1NSAID-regulated gene 1 protein|PDFGDF-15|MIC1Prostate differentiation factor|MIC-1|PTGF-beta|NSAID (nonsteroidal inflammatory drug)-activated protein 1|Placental bone morphogenetic protein|PTGFBPTGF-beta|GDF15|Macrophage inhibitory cytokine 1|NAG-1|PDF|MIC-1NSAID-activated gene 1 protein|PLAB|Placental TGF-beta
  • Accession No.:
    Q99988
  • Protein/Peptide Name:
    GDF-15
  • Purity:
    >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
  • Molecular Weight:
    12-14 kDa, reducing conditions
  • Cat. No.:
    102869-342
  • Supplier no.:
    8146-GD-025/CF

Specifications

About this item

The Recombinant Human GDF-15 (Human Cell-expressed) Protein from R&D Systems is derived from HEK293. The Recombinant Human GDF-15 (Human Cell-expressed) Protein has been validated for the following applications: Bioactivity.