Order Entry
United States
ContactUsLinkComponent
Human Recombinant Fucosyltransferase 2/FUT2 (from HEK293 Cells)
Human Recombinant Fucosyltransferase 2/FUT2 (from HEK293 Cells)
Catalog # 102869-102
Supplier:  R&D Systems
CAS Number:  
Human Recombinant Fucosyltransferase 2/FUT2 (from HEK293 Cells)
Catalog # 102869-102
Supplier:  R&D Systems
Supplier Number:  7770-GT-020
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    20 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    2524
  • Protein Synonyms:
    Fucosyltransferase 2|2)FT2|Alpha(1|SEC2|SEC2SE|FUT2|SE2|alpha(1|EC 2.4.1.69|galactoside 2-alpha-L-fucosyltransferase 2|GDP-L-fucose:beta-D-galactoside 2-alpha-L-fucosyltransferase 2|Secretor factor|sej|fucosyltransferase 2 (secretor status included)|Se|alpha (1|2) fucosyltransferase|B12QTL1|Secretor blood group alpha-2-fucosyltransferase|2)FT 2
  • Accession No.:
    Q10981
  • Protein/Peptide Name:
    Fucosyltransferase 2/FUT2
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
  • Molecular Weight:
    40-50 kDa, reducing conditions
  • Cat. No.:
    102869-102
  • Supplier no.:
    7770-GT-020

Specifications

About this item

The Recombinant Human Fucosyltransferase 2/FUT2 Protein from R&D Systems is derived from HEK293. The Recombinant Human Fucosyltransferase 2/FUT2 Protein has been validated for the following applications: Enzyme Activity.