Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:25 μg
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:10673
- Protein Synonyms:B lymphocyte stimulator|member 13b|TNF and ApoL-related leukocyte expressed ligand 1|CD257 antigen|CD257|tumor necrosis factor superfamily|TALL1Delta4 BAFF|ZTNF4|THANK|tumor necrosis factor ligand superfamily member 13B|Dendritic cell-derived TNF-like molecule|member 20|tumor necrosis factor (ligand) superfamily|TNF- and APOL-related leukocyte expressed ligand 1|TNF homolog that activates apoptosis|TALL-1delta BAFF|TNFSF20|ApoL related ligand TALL-1|BLyS|DTL|BLYSB-lymphocyte stimulator|B-cell-activating factor|TNFSF13B|TALL1|BAFFB-cell activating factor|tumor necrosis factor-like protein ZTNF4
- Accession No.:Q9Y275
- Protein/Peptide Name:BAFF/BLyS/TNFSF13B
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular Weight:19 kDa, reducing conditions
- Cat. No.:102877-234
- Supplier no.:7537-BF-025
The Recombinant Human BAFF/BLyS/TNFSF13B (CHO-expressed) Protein from R&D Systems is derived from CHO. The Recombinant Human BAFF/BLyS/TNFSF13B (CHO-expressed) Protein has been validated for the following applications: Bioactivity.