Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:10 μg
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin Content:<0.01 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:27179
- Protein Synonyms:FIL1 epsilon|FIL1E|FIL1|IL36A|epsilon|member 6 (epsilon)|IL-1 epsilon|Alpha|interleukin 1 family|IL1F6|Interleukin 36|IL1(EPSILON)|Interleukin-36 Alpha|FIL1(EPSILON)|interleukin 1|IL-1F6|Interleukin-1 epsilon|IL-1E|IL36 alpha|IL-1F6 (FIL-1-epsilon)|interleukin-1 family member 6|IL1E
- Accession No.:NP_055255
- Protein/Peptide Name:IL-36 alpha/IL-1F6
- Purity:>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
- Molecular Weight:16 kDa, reducing conditions
- Cat. No.:102867-642
- Supplier no.:6995-IL-010/CF
The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein from R&D Systems is derived from E. coli. The Recombinant Human IL-36 alpha/IL-1F6 (aa 6-158) Protein has been validated for the following applications: Bioactivity.