To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:20 μg
- Environmentally Preferable:
- Storage Conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening.
- Endotoxin Content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:28
- Protein Synonyms:A transferase|Histo-blood group A transferase|A3GALNT|Fucosylglycoprotein 3-alpha-galactosyltransferase|histo-blood group A2 transferase|Histo-blood group B transferase|NAGAT|Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase|EC 2.4.1.37|Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase|GTB|B transferase|histo-blood group ABO system transferase|A3GALT1|ABO blood group (transferase A|ABO glycosyltransferase|ABO|alpha 1-3-galactosyltransferase)|alpha 1-3-N-acetylgalactosaminyltransferase|Glycoprotein-fucosylgalactoside alpha-galactosyltransferase|transferase B|B(A) alpha-1,3-galactosyltransferase|EC 2.4.1.40
- Accession No.:AAD26574
- Protein/Peptide Name:Blood Group B Transferase/GTB/ABO
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
- Molecular Weight:35-40 kDa, reducing conditions
- Cat. No.:102868-688
- Supplier no.:6824-GT-020
The Recombinant Human Blood Group B Transferase/GTB Protein from R&D Systems is derived from CHO. The Recombinant Human Blood Group B Transferase/GTB Protein has been validated for the following applications: Enzyme Activity.