Order Entry
United States
ContactUsLinkComponent
Human Recombinant PLK1 (from Baculovirus (Sf9 Insect Cells))
Human Recombinant PLK1 (from Baculovirus (Sf9 Insect Cells))
Catalog # 220009-382
Supplier:  R&D Systems
CAS Number:  
Human Recombinant PLK1 (from Baculovirus (Sf9 Insect Cells))
Catalog # 220009-382
Supplier:  R&D Systems
Supplier Number:  3804-KS-010
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Baculovirus (Sf9 Insect cells)
  • Species:
    Human
  • Size:
    10 μg
  • Storage Conditions:
    This product is stable at <= -70░ C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles.
  • Gene ID:
    5347
  • Protein Synonyms:
    STPK13|serine/threonine-protein kinase PLK1|EC 2.7.11.21|Serine/threonine-protein kinase 13|PLK|polo (Drosophia)-like kinase|cell cycle regulated protein kinase|polo like kinase|polo-like kinase 1polo-like kinase (Drosophila)|EC 2.7.11|PLK-1
  • Accession No.:
    NM_005030
  • Protein/Peptide Name:
    PLK1
  • Purity:
    >70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
  • Molecular Weight:
    70 kDa
  • Cat. No.:
    220009-382
  • Supplier no.:
    3804-KS-010

Specifications

About this item

The Recombinant Human Active PLK1 Protein from R&D Systems is derived from Sf 9 (baculovirus). The Recombinant Human Active PLK1 Protein has been validated for the following applications: Bioactivity.