Order Entry
United States
Orders LinkContactUsLinkComponent
Human Coagulation Factor II/Thrombin (from Plasma)

Human Coagulation Factor II/Thrombin (from Plasma)

200 μg
Catalog # 220023-473
Marketsource
Supplier:   R&D Systems
Supplier #:  2196-SE-200
Human Coagulation Factor II/Thrombin (from Plasma)
Catalog # 220023-473
Supplier:  R&D Systems
Marketsource
CAS Number:  
Human Coagulation Factor II/Thrombin (from Plasma)

200 μg

CAS Number:  
Price
$308.03
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

The Human Coagulation Factor II/Thrombin Protein from R&D Systems is derived from Human Plasma. The Human Coagulation Factor II/Thrombin Protein has been validated for the following applications: Enzyme Activity.

Specifications

Specifications

  • Size:
    200 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Cat. No.:
    220023-473
  • Molecular Weight:
    83 kDa, reducing conditions
  • Gene ID:
    2147
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Protein Synonyms:
    Coagulation factor II receptor-like 2|Coagulation factor II receptor-like 2 (protease-actovated receptor 3)|PT|Thrombin receptor-like 2|PAR3proteinase-activated receptor 3|proteinase-activated receptor-3|serine protease|protease-activated receptor 3|Thrombin|coagulation factor II (thrombin) receptor-like 2|Coagulation Factor II|F2|PAR-3
  • Protein/Peptide Name:
    Coagulation Factor II/Thrombin
  • Conjugation:
    Unconjugated
  • Endotoxin Content:
    <1.0 EU per 1 ╡g of the protein by the LAL method.
  • Species:
    Human
  • Source:
    Plasma

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.