Order Entry
United States
ContactUsLinkComponent
Rat Recombinant MIS RII/AMHR2 (from Insect Cells (Sf21))
Rat Recombinant MIS RII/AMHR2 (from Insect Cells (Sf21))
Catalog # 220023-103
Supplier:  R&D Systems
CAS Number:  
Rat Recombinant MIS RII/AMHR2 (from Insect Cells (Sf21))
Catalog # 220023-103
Supplier:  R&D Systems
Supplier Number:  1618-MR-050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    Insect cells (Sf21)
  • Species:
    Rat
  • Size:
    50 μg
  • Storage Conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin Content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    269
  • Protein Synonyms:
    Muellerian hormone type II receptor|Mullerian hormone receptor|Mullerian inhibiting substance type II receptor|C14|AMHRII|MISRIIMIS type II receptor|type II|MISR2|MISRII|Muellerian hormone type-2 receptor|AMH type II receptor|MRII|AMHR2|AMHREC 2.7.11.30
  • Accession No.:
    Q62893
  • Protein/Peptide Name:
    MIS RII/AMHR2
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Molecular Weight:
    50-55 kDa, reducing conditions
  • Cat. No.:
    220023-103
  • Supplier no.:
    1618-MR-050

Specifications

About this item

The Recombinant Rat MIS RII Fc Chimera Protein from R&D Systems is derived from Sf 21 (stably transfected). The Recombinant Rat MIS RII Fc Chimera Protein has been validated for the following applications: Binding Activity.