BMS-345541 free base ≥98% (by HPLC)
Supplier: Avantor
Certificates
About this item
Synonyms: N1-(1,8-Dimethylimidazo[1,2-a]quinoxalin-4-yl)ethane-1,2-diamine, N-(1,8-Dimethylimidazo[1,2-a]quinoxalin-4-yl)-1,2-ethanediamine
CAS Number:445430-58-0
:MFCD07784499
Formula:C₁₄H₁₇N₅
MDL Number:MFCD18089812
Molecular Weight:255.32 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together