Anti-M-CSF Rabbit Polyclonal Antibody (Biotin)
Supplier: Peprotech
Certificates
About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human M-CSF. AntiHuman M-CSFspecific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: M-CSF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Specifications
- Catalog No:
- 10775-334
- 10775-332
- Antigen Symbol:
- M-CSF
- M-CSF
- Antigen Name:
- M-CSF
- M-CSF
- Antibody Type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human M-CSF
- Recombinant Human M-CSF
- Form:
- Lyophilized
- Lyophilized
- Molecular Weight:
- 36.8 kDa
- 36.8 kDa
- Purification:
- Recombinant Human M-CSF
- Recombinant Human M-CSF
- Storage Buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage Temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together