About this item
Produced from sera of rabbits immunized with highly pure Recombinant Murine IP-10 (CXCL10). AntiMurine IP-10 (CXCL10)specific antibody was purified by affinity chromatography employing an immobilized Murine IP-10 matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: IP-10 (CXCL10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10776-254
- 10776-252
- 10776-250
- Antigen Symbol:
- IP-10 (CXCL10)
- IP-10 (CXCL10)
- IP-10 (CXCL10)
- Antigen Name:
- IP-10
- IP-10
- IP-10
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Murine IP-10 (CXCL10)
- Recombinant Murine IP-10 (CXCL10)
- Recombinant Murine IP-10 (CXCL10)
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular Weight:
- 8.7 kDa
- 8.7 kDa
- 8.7 kDa
- Purification:
- Recombinant Murine IP-10 (CXCL10)
- Recombinant Murine IP-10 (CXCL10)
- Recombinant Murine IP-10 (CXCL10)
- Storage Buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage Temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping Temperature:
- Ambient
- Ambient
- Ambient