Order Entry
United States
Orders LinkContactUsLinkComponent
Anti-FGF-basic Rabbit Polyclonal Antibody

Anti-FGF-basic Rabbit Polyclonal Antibody

Supplier:   Peprotech
Anti-FGF-basic Rabbit Polyclonal Antibody
Supplier:  Peprotech
Certificates

3 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-basic. Anti­Human FGF-basic­specific antibody was purified by affinity chromatography employing an immobilized Human FGF-basic matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: FGF-basic
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human

Specifications for Product Family

Specifications

  • Catalog No:
  • 10775-082
  • 10775-084
  • 10775-086
  • Antigen Symbol:
  • FGF-basic
  • FGF-basic
  • FGF-basic
  • Antigen Name:
  • FGF-basic
  • FGF-basic
  • FGF-basic
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western Blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human FGF-basic
  • Recombinant Human FGF-basic
  • Recombinant Human FGF-basic
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Molecular Weight:
  • 17.2 kDa
  • 17.2 kDa
  • 17.2 kDa
  • Purification:
  • Recombinant Human FGF-basic
  • Recombinant Human FGF-basic
  • Recombinant Human FGF-basic
  • Storage Buffer:
  • See Product Data Sheet for more information.
  • See Product Data Sheet for more information.
  • See Product Data Sheet for more information.
  • Storage Temperature:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping Temperature:
  • Ambient
  • Ambient
  • Ambient

Specifications

Product Family Options

Product Information

  • Size
    Packaging
    Availability
    Price
  • 50 µg
    PP Vial
    Log in for availability
    Log in for availability
    Each (50µG)
    Each (50µG) - $390.37
    $390.37
    Each (50µG)
       $390.37
    Minimum order quantity is 1

    Supplier Number: 500-P18-50UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10775-082
    Antigen Symbol
    FGF-basic
    Antigen Name
    FGF-basic
    Antibody Type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Isotype
    IgG
    Western Blot
    Yes
    ELISA
    Yes
    Immunogen
    Recombinant Human FGF-basic
    Form
    Lyophilized
    Molecular Weight
    17.2 kDa
    Purification
    Recombinant Human FGF-basic
    Storage Buffer
    See Product Data Sheet for more information.
    Storage Temperature
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 100 µg
    PP Vial
    Log in for availability
    Log in for availability
    Each (100µG)
    Each (100µG) - $518.20
    $518.20
    Each (100µG)
       $518.20
    Minimum order quantity is 1

    Supplier Number: 500-P18-100UG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10775-084
    Antigen Symbol
    FGF-basic
    Antigen Name
    FGF-basic
    Antibody Type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Isotype
    IgG
    Western Blot
    Yes
    ELISA
    Yes
    Immunogen
    Recombinant Human FGF-basic
    Form
    Lyophilized
    Molecular Weight
    17.2 kDa
    Purification
    Recombinant Human FGF-basic
    Storage Buffer
    See Product Data Sheet for more information.
    Storage Temperature
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient
  • 1 mg
    PP Vial
    Log in for availability
    Log in for availability
    Each (1mg)
    Each (1mg) - $3,028.31
    $3,028.31
    Each (1mg)
       $3,028.31
    Minimum order quantity is 1

    Supplier Number: 500-P18-1MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    10775-086
    Antigen Symbol
    FGF-basic
    Antigen Name
    FGF-basic
    Antibody Type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Isotype
    IgG
    Western Blot
    Yes
    ELISA
    Yes
    Immunogen
    Recombinant Human FGF-basic
    Form
    Lyophilized
    Molecular Weight
    17.2 kDa
    Purification
    Recombinant Human FGF-basic
    Storage Buffer
    See Product Data Sheet for more information.
    Storage Temperature
    Store at –20 °C prior to reconstitution.
    Shipping Temperature
    Ambient