Anti-MIP-3alpha (CCL20) Mouse Monoclonal Antibody
Catalog # 10774-858
Supplier: Peprotech
Specifications
- Antibody Type:Primary
- Antigen Name:MIP-3alpha
- Antigen Symbol:CCL20
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:500 µg
- Form:Lyophilized
- Storage Buffer:See Product Data Sheet for more information.
- Molecular Weight:8.0 kDa
- Storage Temperature:Store at –20 °C prior to reconstitution.
- Shipping Temperature:Ambient
- Immunogen:Recombinant Human MIP-3α (CCL20)
- Purification:Recombinant Human MIP-3α (CCL20)
- Cat. No.:10774-858
- Packaging:PP Vial
- Supplier no.:500M28500UG
Specifications
About this item
Monoclonal AntibodyProduced in mice immunized with highly pure Recombinant Human MIP-3α (CCL20). AntiHuman MIP-3α (CCL20)specific antibody was purified from cell culture by Protein A affinity chromatography.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: MIP-3alpha (CCL20)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human