To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:MCP-2
- Antigen Symbol:MCP-2 (CCL8)
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:500 µg
- Form:Lyophilized
- Storage Buffer:See Product Data Sheet for more information.
- Molecular Weight:8.9 kDa
- Storage Temperature:Store at –20 °C prior to reconstitution.
- Shipping Temperature:Ambient
- Immunogen:Recombinant Human MCP-2 (CCL8)
- Purification:Recombinant Human MCP-2 (CCL8)
- Cat. No.:10774-894
- Packaging:PP Vial
- Supplier no.:500M69500UG
Specifications
About this item
Monoclonal AntibodyProduced in BALB/c x ICR F1-mice immunized with highly pure Recombinant Human MCP-2 (CCL8). AntiHuman MCP-2 (CCL8)specific antibody was purified from ascites fluid by Protein A affinity chromatography.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: MCP-2 (CCL8)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human