Order Entry
United States
ContactUsLinkComponent
Anti-IP-10 (CXCL10) Mouse Monoclonal Antibody
Anti-IP-10 (CXCL10) Mouse Monoclonal Antibody
Catalog # 10774-888
Supplier:  Peprotech
Anti-IP-10 (CXCL10) Mouse Monoclonal Antibody
Catalog # 10774-888
Supplier:  Peprotech
Supplier Number:  500-M60-500UG

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    IP-10
  • Antigen Symbol:
    IP-10 (CXCL10)
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    500 µg
  • Form:
    Lyophilized
  • Storage Buffer:
    See Product Data Sheet for more information.
  • Molecular Weight:
    8.6 kDa
  • Storage Temperature:
    Store at –20 °C prior to reconstitution.
  • Shipping Temperature:
    Ambient
  • Immunogen:
    Recombinant Human IP-10 (CXCL10)
  • Purification:
    Recombinant Human IP-10 (CXCL10)
  • Cat. No.:
    10774-888
  • Packaging:
    PP Vial
  • Supplier no.:
    500M60500UG

Specifications

About this item

Monoclonal AntibodyProduced in BALB/c-mice immunized with highly pure Recombinant Human IP-10 (CXCL10). Anti­Human IP-10 (CXCL10)­specific antibody was purified from cell culture by Protein A affinity chromatography.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IP-10 (CXCL10)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human