Rabbit IgG Isotype Control
500 µg, PP Vial
Catalog # 10774-932
Supplier: Peprotech
Supplier #: 500-P00-500UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Monoclonal AntibodyNormal rabbit IgG is produced from serum of rabbits that have not been immunized. This IgG antibody was purified by Protein A chromatography. This preparation can be used as a control reagent for immunoasssays using rabbit polyclonal antibodies.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Control
Antigen:
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Size:500 µg
- Packaging:PP Vial
- Cat. No.:10774-932
- Shipping Temperature:Ambient
- Antibody Type:Control
- Form:Lyophilized
- Storage Buffer:See Product Data Sheet for more information.
- Host:Rabbit
- Conjugation:Unconjugated
- Storage Temperature:Store at –20 °C prior to reconstitution.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



