ContactUsLinkComponent
Trypkit™ Xeno-Free Large, Lifeline Cell Technology
Trypkit™ Xeno-Free Large, Lifeline Cell Technology
Catalog # 102970-896
CAS Number:  
Trypkit™ Xeno-Free Large, Lifeline Cell Technology
Catalog # 102970-896
Supplier Number:  LL-0043
CAS Number:  
Specifications
  • Description:
    TrypKit™, Xeno-Free - Large Kit
  • Size:
    100 ml
  • Cat. No.:
    102970-896
  • Supplier no.:
    LL-0043
Specifications
About this item
Delivery: The kit includes PBS XF (500 ml), trypsin EDTA XF (100 ml) and trypsin neutralizing solution XF (100 ml).