ContactUsLinkComponent
Ferric Reducing Antioxidant Power (FRAP) Assay Kit (Colorimetric), BioVision, Inc.
Ferric Reducing Antioxidant Power (FRAP) Assay Kit (Colorimetric), BioVision, Inc.
Catalog # 76200-036
Supplier:  Avantor
CAS Number:  
undefined
Ferric Reducing Antioxidant Power (FRAP) Assay Kit (Colorimetric), BioVision, Inc.
Catalog # 76200-036
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Ferric Reducing Antioxidant Power (FRAP) Assay Kit (Colorimetric)
  • Size:
    200 assays
  • Cat. No.:
    76200-036
Specifications
About this item
Convenient and reliable assay to measure total antioxidant capacity from serum, urine, and foodstuff.
  • Simple procedure, takes less than 2 hours
  • Fast and convenient
Ferric reducing antioxidant power (FRAP) assay is a widely used method that uses antioxidants as reductants in a redox-linked colorimetric reaction, wherein Fe3+ is reduced to Fe2+. Ferric (Fe3+) to ferrous (Fe2+) ion reduction at low pH causes formation of a colored ferrous-probe complex from a colorless ferric-probe complex.
Antioxidants are molecules which act as reducing agents by donating electrons to free radicals to stabilize them and minimize the damage caused by free radicals to DNA, cells and organ systems. Antioxidants include substances such as polyphenols; flavonoids, vitamins and enzymes like glutathione peroxidase and superoxide dismutase. They are known to have beneficial health effects such as lowering the risk of cancer, heart disease and neurodegenerative disorders and are abundantly found in plants, fruits, vegetables, beverages and natural products.
The kit provides a quick, sensitive and easy way for measuring antioxidant capacity of various biological samples. The assay is high-throughput adaptable and can detect antioxidant capacities as low as 0.2 mM Fe2+ equivalents.