Human IL-33 Standard ABTS ELISA Development Kit, PeproTech, Inc.
PeproTech's Human IL-33 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-33 in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human IL-33 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-33 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human IL-33 in approximately 1000 ELISA plate wells.
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:IL-33
Frequently Bought Together