About this item
PeproTech's Human TNF-α ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human TNF-α in a sandwich ELISA format.
Human TNF-α ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human TNF-α in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human TNF-α approximately 1000 ELISA plate wells.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:TNF-α