Order Entry
United States
ContactUsLinkComponent
Murine MDC (CCL22) Standard ABTS ELISA Development Kit, PeproTech
Murine MDC (CCL22) Standard ABTS ELISA Development Kit, PeproTech
Catalog # 10771-600
Supplier:  Peprotech
CAS Number:  
Murine MDC (CCL22) Standard ABTS ELISA Development Kit, PeproTech
Catalog # 10771-600
Supplier:  Peprotech
Supplier Number:  900-K197
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Rat
    Mouse
  • Description:
    Murine MDC (CCL22) ELISA Kit
  • Size:
    Kit components sufficient to assay 10 plates
  • Cat. No.:
    10771-600
  • Supplier no.:
    900-K197

Specifications

About this item

PeproTech's Murine MDC (CCL22) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine MDC (CCL22) in a sandwich ELISA format.

  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)

Murine MDC (CCL22) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine MDC (CCL22) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay murine MDC (CCL22) in approximately 1000 ELISA plate wells.