Human FGF-basic Standard ABTS ELISA Development Kit, PeproTech, Inc.
About this item
PeproTech's Human FGF-basic ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human FGF-basic in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human FGF-basic ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human FGF-basic in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human FGF-basic approximately 1000 ELISA plate wells.
1 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
PeproTech's Human FGF-basic ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human FGF-basic in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human FGF-basic ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human FGF-basic in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human FGF-basic approximately 1000 ELISA plate wells.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10777-634
- 10777-772
- Primary antibody reactivity:
- Human
- Human
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 10777-634Human FGF-basic ELISA KitHuman FGF-basic ELISA KitKit components sufficient to assay 10 platesLog in for availabilityKit of 10$659.20Supplier Number: 900-K08KItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10777-634Primary antibody reactivityHuman - 10777-772Human FGF-basic Mini-ELISA KitHuman FGF-basic Mini-ELISA KitKit components sufficient to assay 2 platesDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10777-772Primary antibody reactivityHuman