Order Entry
United States
Orders LinkContactUsLinkComponent
ABTS ELISA Buffer Kit, PeproTech, Inc.

ABTS ELISA Buffer Kit, PeproTech, Inc.

ELISA Buffer Kit, Kit components sufficient to assay 10 plates
Catalog # 10777-628
Supplier:   Peprotech
Supplier #:  900-K00
ABTS ELISA Buffer Kit, PeproTech, Inc.
Catalog # 10777-628
Supplier:  Peprotech
CAS Number:  
ABTS ELISA Buffer Kit, PeproTech, Inc.

ELISA Buffer Kit, Kit components sufficient to assay 10 plates

CAS Number:  
Price
$428.07
Log in for availability
Kit of 10
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

PeproTech's ABTS ELISA Buffer Kit is specifically formulated to be used in conjunction with PeproTech’s Standard ABTS ELISA Development Kits.

  • Sterile-filtered buffers
  • Ready-to-use ABTS substrate
  • Flexibility: Use with any ELISA Kit
  • Uncoated plates

The buffers in this supplemental kit have been specially formulated for optimal performance when used in conjunction with PeproTech’s ELISA Development Kits. The buffer kit contains all of the necessary components to assay ten 96-well ELISA plates (included) and contains detailed handling instructions. All of the reagents have been sterile-filtered to minimize assay interference and maximize shelf life. This easy-to-use ELISA buffer kit can also be purchased as a stand-alone product since the plates included are not pre-coated with capture antibody. This format allows the researcher to develop and optimize an assay for use with their own capture and detection antibodies. The actual antibody concentrations and detection ranges of the ELISA will vary.

Kit Contents:
- 20X Plate Coating Buffer (PBS)
- 1X Blocking Buffer
- 20X Sample Diluent
- 20X Wash Buffer
- Ready-to-use ABTS
- 10 Sterile, Uncoated 96-well ELISA Plates
- 50 Plate Sealing Films
- Buffer Handling Instructions

Specifications

Specifications

  • Description:
    ELISA Buffer Kit
  • Size:
    Kit components sufficient to assay 10 plates
  • Cat. No.:
    10777-628
  • Target protein:
    ABTS

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.