ContactUsLinkComponent
PicoProbe™ Reduced Glutathione (GSH) Assay Kit (Fluorometric), BioVision
PicoProbe™ Reduced Glutathione (GSH) Assay Kit (Fluorometric), BioVision
Catalog # 76200-060
Supplier:  Avantor
CAS Number:  
undefined
PicoProbe™ Reduced Glutathione (GSH) Assay Kit (Fluorometric), BioVision
Catalog # 76200-060
Supplier:  Avantor
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    PicoProbe™ Reduced Glutathione (GSH) Assay Kit (Fluorometric)
  • Size:
    100 assays
  • Cat. No.:
    76200-060
Specifications
About this item
It specifically detects reduced glutathione. Oxidized GSSG is not detected.
  • Simple procedure, takes only approximately 1 to 2 hours
  • Fast and convenient
  • Specific: does not detect oxidized GSSG
  • Sensitive: detects as low as 8 pmol GSH
Glutathione (GSH), a thiol-containing tripeptide, is a key antioxidant in living systems. Intracellular GSH status appears to be a sensitive indicator of the overall health of a cell. THe kit is based on a specific enzymatic cycling method in the presence of GSH and a fluorophore.
The reduction of the fluorophore produces a stable fluorescent product, the fluorescence of which is directly proportional to the amount of GSH in the sample and can be followed kinetically (Ex/Em = 535/587 nm). Our assay is the most specific and sensitive in the market. Oxidized Glutathione does not interfere with the assay. The assay is simple, reproducible and can specifically detect as low as 8 pmol/well of reduced form of Glutathione (GSH) in a 100 µl reaction